Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100910678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749743.3 | XP_003749791.1 | 2 | 166 | ||
XM_006236476.2 | XP_006236538.1 | 1 | 168 |
Gene Symbol: Tma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126048.2 | NP_001119520.1 | 3 | 190 | ||
GenBank mRNA | BC157813.1 | - | 3 | 190 | ||
FQ221914.1 | - | 3 | 166 | |||
FQ222972.1 | - | 3 | 155 |