Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024351.2 | NP_077327.1 | 8 | 1704 | ||
GenBank mRNA | BC061547.1 | - | 8 | 1711 | 6921606 | |
BC098914.1 | - | 8 | 1656 | 7104230 | ||
FQ213866.1 | - | 4 | 720 | |||
FQ226406.1 | - | 4 | 639 | |||
FQ231618.1 | - | 3 | 351 | |||
FQ231904.1 | - | 4 | 896 | |||
FQ232261.1 | - | 4 | 639 | |||
FQ234652.1 | - | 4 | 1053 | |||
FQ235115.1 | - | 4 | 1053 | |||
M11942.1 | - | 8 | 1675 |
Gene Symbol: LOC689955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764620.1 | XP_008762842.1 | 1 | 1706 |