Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TP53I11 SIRNA Assays ›
Gene Symbol:
TP53I11
Gene Name:
tumor protein p53 inducible protein 11
Gene Aliases:
PIG11
Chromosome Location:
Chr.11: 44932348 - 44951306 on Build GRCh38
UniGene ID:
Hs.554791
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18310
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TP53I11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258320.1
NP_001245249.1
6
811
NM_001258321.1
NP_001245250.1
4
735
NM_001258322.1
NP_001245251.1
4
665
NM_001258323.2
NP_001245252.1
4
492
NM_001258324.1
NP_001245253.1
3
619
NM_001318384.1
NP_001305313.1
4
500
NM_001318385.1
NP_001305314.1
4
696
NM_001318386.1
NP_001305315.1
4
730
NM_001318389.1
NP_001305318.1
3
417
NM_001318390.1
NP_001305319.1
3
417
NM_006034.4
NP_006025.2
3
417
NR_134612.1
3
417
XM_005253227.4
XP_005253284.1
4
512
XM_005253229.1
XP_005253286.1
3
417
XM_011520475.2
XP_011518777.1
4
520
XM_011520476.2
XP_011518778.1
4
541
XM_011520477.2
XP_011518779.1
4
564
XM_011520478.2
XP_011518780.1
4
473
XM_017018580.1
XP_016874069.1
4
380
XM_017018581.1
XP_016874070.1
3
214
GenBank mRNA
AK026607.1
-
3
391
AK054815.1
-
4
471
AK056379.1
-
4
428
AK292837.1
-
3
229
AL537510.3
-
3
370
BC045666.1
-
6
763
5298525
BU149365.1
-
366
DA115495.1
-
4
280
DA120301.1
-
4
323
DA410127.1
-
4
353
DA697978.1
-
4
350
DA729260.1
-
3
353
DA770050.1
-
4
541
DA778197.1
-
4
364
DA801070.1
-
3
355
DB166321.1
-
3
353
U79302.1
-
3
602
Back To Top
More Information
Gene Ontology Categories:
Function(s)
response to stress
negative regulation of cell proliferation
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0