Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tirap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001055833.4 | XP_001055833.2 | 2 | 435 | ||
XM_006242794.2 | XP_006242856.1 | 2 | 435 |