Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVPR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000706.4 | NP_000697.1 | 1, 2 | 2942 | ||
GenBank mRNA | AF030625.1 | - | 1, 2 | 991 | ||
AF101725.1 | - | 1, 2 | 991 | |||
AY322550.1 | - | 1, 2 | 968 | |||
BC074803.2 | - | 1, 2 | 991 | 30915313 | ||
BC074804.2 | - | 1, 2 | 991 | 30915527 | ||
KJ534779.1 | - | 1, 2 | 968 | |||
KJ534780.1 | - | 2, 3 | 860 | |||
L25615.1 | - | 1, 2 | 1020 | |||
S73899.1 | - | 1, 2 | 1084 |
Set Membership: |
![]() |