Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005458.7 | NP_005449.5 | 7 | 1483 | ||
XM_005252316.4 | XP_005252373.1 | 5 | 516 | |||
XM_017015331.1 | XP_016870820.1 | 6 | 901 | |||
XM_017015332.1 | XP_016870821.1 | 4 | 470 | |||
GenBank mRNA | AF056085.1 | - | 7 | 1480 | ||
AF069755.1 | - | 7 | 1179 | |||
AF074483.1 | - | 7 | 1023 | |||
AF095784.1 | - | 7 | 1154 | |||
AF099033.1 | - | 7 | 1023 | |||
AJ012188.1 | - | 7 | 1023 | |||
AK308139.1 | - | 6 | 656 | |||
BC035071.2 | - | 8 | 1318 | 5260138 | ||
JQ308173.1 | - | 7 | 1023 |
Set Membership: |
![]() |