Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005458.7 | NP_005449.5 | 4 | 1167 | ||
XM_005252316.4 | XP_005252373.1 | 2 | 200 | |||
XM_017015331.1 | XP_016870820.1 | 3 | 585 | |||
GenBank mRNA | AF056085.1 | - | 4 | 1164 | ||
AF069755.1 | - | 4 | 863 | |||
AF074483.1 | - | 4 | 707 | |||
AF095784.1 | - | 4 | 838 | |||
AF099033.1 | - | 4 | 707 | |||
AJ012188.1 | - | 4 | 707 | |||
AK308139.1 | - | 3 | 340 | |||
BC035071.2 | - | 5 | 1002 | 5260138 | ||
JQ308173.1 | - | 4 | 707 |
Set Membership: |
![]() |