Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253854.1 | NP_001240783.1 | 6, 7 | 813 | ||
NM_001253855.1 | NP_001240784.1 | 2, 3 | 544 | |||
NM_012443.3 | NP_036575.1 | 3, 4 | 458 | |||
NM_172242.2 | NP_758442.1 | 3, 4 | 458 | |||
XM_005252645.1 | XP_005252702.1 | 3, 4 | 487 | |||
XM_005252646.1 | XP_005252703.1 | 3, 4 | 487 | |||
GenBank mRNA | AF079363.1 | - | 3, 4 | 415 | ||
AK289903.1 | - | 3, 4 | 407 | |||
AK302194.1 | - | 6, 7 | 788 | |||
AK310247.1 | - | 2, 3 | 497 | |||
AL080136.1 | - | 3, 4 | 406 | |||
BC030585.2 | - | 3, 4 | 406 | 4822996 | ||
BI561430.1 | - | 3, 4 | 437 | |||
CR533552.1 | - | 3, 4 | 291 | |||
DB025758.1 | - | 3, 4 | 442 | |||
DB057157.1 | - | 3, 4 | 460 |