Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004905.2 | NP_004896.1 | 2 | 268 | ||
GenBank mRNA | AJ844621.1 | - | 2 | 179 | ||
AK289352.1 | - | 2 | 266 | |||
AK300680.1 | - | 2 | 278 | |||
BC035857.1 | - | 2 | 268 | 5767682 | ||
BC053550.1 | - | 2 | 218 | 6013037 | ||
D14662.1 | - | 2 | 260 | |||
EU794652.1 | - | 2 | 268 |