Gene Symbol: IER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004907.2 | NP_004898.2 | 2 | 755 | ||
GenBank mRNA | BC003625.2 | - | 1 | 535 | 2966293 | |
BC072432.1 | - | 2 | 755 | 5727038 | ||
BC110647.1 | - | 1 | 1639 | 4416077 | ||
BT019466.1 | - | 1 | 427 | |||
CR450352.1 | - | 1 | 427 | |||
M62831.1 | - | 1 | 527 |