Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109510.1 | NP_001102980.1 | 2 | 822 |
Set Membership: |
G-protein coupled receptor |