Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085406.1 | NP_001078875.1 | 11 | 1291 | ||
XM_008766773.1 | XP_008764995.1 | 12 | 1579 | |||
GenBank mRNA | BC101893.1 | - | 10 | 1119 | ||
BC158552.1 | - | 11 | 1291 | 9098910 | ||
FQ226713.1 | - | 3 | 392 |