Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191087.1 | NP_001178016.1 | 250 |
Gene Symbol: LOC100909604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762945.1 | XP_008761167.1 | 1 | 4681 |
Gene Symbol: LOC100911511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752179.3 | XP_003752227.1 | 1 | 2626 |