Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGS6 SIRNA Assays ›
Gene Symbol:
RGS6
Gene Name:
regulator of G-protein signaling 6
Gene Aliases:
GAP
Chromosome Location:
Chr.14: 71932100 - 72632478 on Build GRCh38
UniGene ID:
Hs.509872
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18503
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RGS6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204416.2
NP_001191345.1
4, 5
526
NM_001204417.2
NP_001191346.1
4, 5
526
NM_001204418.2
NP_001191347.1
4, 5
526
NM_001204419.2
NP_001191348.1
4, 5
526
NM_001204420.2
NP_001191349.1
4, 5
526
NM_001204421.2
NP_001191350.1
4, 5
526
NM_001204422.2
NP_001191351.1
4, 5
526
NM_001204423.1
NP_001191352.1
3, 4
291
NM_001204424.1
NP_001191353.1
4, 5
764
NM_004296.6
NP_004287.3
4, 5
526
NR_135235.1
4, 5
526
XM_011537393.2
XP_011535695.1
3, 4
775
XM_011537397.1
XP_011535699.1
1, 2
57
XM_011537407.2
XP_011535709.1
3, 4
778
XM_017021818.1
XP_016877307.1
4, 5
426
XM_017021819.1
XP_016877308.1
4, 5
426
XM_017021820.1
XP_016877309.1
3, 4
261
XM_017021821.1
XP_016877310.1
3, 4
261
XM_017021822.1
XP_016877311.1
3, 4
261
XM_017021823.1
XP_016877312.1
3, 4
261
XM_017021824.1
XP_016877313.1
4, 5
426
XM_017021825.1
XP_016877314.1
3, 4
261
XM_017021826.1
XP_016877315.1
3, 4
261
XM_017021827.1
XP_016877316.1
3, 4
261
XM_017021828.1
XP_016877317.1
3, 4
261
XM_017021829.1
XP_016877318.1
4, 5
426
XM_017021830.1
XP_016877319.1
3, 4
261
XM_017021831.1
XP_016877320.1
3, 4
261
XM_017021832.1
XP_016877321.1
3, 4
261
XM_017021833.1
XP_016877322.1
3, 4
261
XM_017021834.1
XP_016877323.1
1, 2
57
GenBank mRNA
AB209462.1
-
4, 5
332
AF073920.2
-
4, 5
448
AF107619.1
-
3, 4
241
AF107620.1
-
3, 4
241
AF156932.1
-
3, 4
241
AK294486.1
-
4, 5
764
AK294895.1
-
3, 4
291
AK302319.1
-
2, 3
137
AK310463.1
-
3, 4
400
AK313481.1
-
4, 5
586
AY309097.1
-
3, 4
241
AY309098.1
-
3, 4
241
AY309099.1
-
3, 4
241
AY309100.1
-
3, 4
241
BC104877.1
-
4, 5
419
8143880
BC113572.1
-
4, 5
419
8322624
BC143699.1
-
4, 5
419
DC317315.1
-
526
DC343081.1
-
495
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
negative regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
signal transducer activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-8tvtn:80/10.239.165.222:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0