Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115035.1 | NP_001108507.1 | 696 | |||
GenBank mRNA | BC158748.1 | - | 664 | 9101450 | ||
BC158821.1 | - | 692 | 7375325 | |||
FQ209543.1 | - | 678 | ||||
FQ213040.1 | - | 677 | ||||
FQ220584.1 | - | 680 | ||||
FQ220807.1 | - | 681 | ||||
FQ230070.1 | - | 682 | ||||
FQ231084.1 | - | 672 | ||||
FQ231314.1 | - | 672 |