Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126299.2 | NP_001119771.2 | 7 | 2578 | ||
XM_006254412.2 | XP_006254474.1 | 9 | 3191 | |||
XM_006254413.2 | XP_006254475.1 | 9 | 3001 | |||
XM_006254414.2 | XP_006254476.1 | 9 | 3188 | |||
GenBank mRNA | BC166441.1 | - | 7 | 2578 | 7461161 |