Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025009.4 | NP_079285.2 | 19 | 2737 | ||
XM_005265788.3 | XP_005265845.1 | 19 | 2704 | |||
XM_006714055.2 | XP_006714118.1 | 19 | 2704 | |||
XM_011534412.2 | XP_011532714.1 | 9 | 1056 | |||
GenBank mRNA | AB014535.1 | - | 12 | 2390 | ||
BC136535.1 | - | 19 | 2564 | 9020525 | ||
BC136536.1 | - | 19 | 2564 | 9020526 |