Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172063.2 | NP_001165534.1 | 8, 9 | 1598 | ||
XM_006242294.2 | XP_006242356.1 | 10, 11 | 2684 | |||
XM_006242295.2 | XP_006242357.1 | 10, 11 | 2007 | |||
XM_006242296.2 | XP_006242358.1 | 10, 11 | 1951 | |||
XM_008765651.1 | XP_008763873.1 | 11, 12 | 2186 | |||
XM_008765652.1 | XP_008763874.1 | 10, 11 | 1943 | |||
XM_008765653.1 | XP_008763875.1 | 10, 11 | 2223 | |||
GenBank mRNA | FQ211776.1 | - | 10, 11 | 1975 |