Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238557.2 | XP_006238619.1 | 4 | 415 | ||
XM_008763931.1 | XP_008762153.1 | 4 | 415 | |||
XM_008763932.1 | XP_008762154.1 | 4 | 405 | |||
XM_008763933.1 | XP_008762155.1 | 4 | 724 | |||
XM_008763934.1 | XP_008762156.1 | 5 | 539 |