Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001071392.5 | XP_001071392.2 | 2 | 1396 | ||
XM_006237431.2 | XP_006237493.1 | 2 | 1398 |