Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748890.3 | XP_003748938.1 | 3 | 1393 | ||
XM_006229465.2 | XP_006229527.1 | 3 | 1396 | |||
XM_006229466.2 | XP_006229528.1 | 3 | 1481 | |||
XM_006229467.2 | XP_006229529.1 | 3 | 1286 | |||
XM_008759592.1 | XP_008757814.1 | 3 | 1283 | |||
XM_008759593.1 | XP_008757815.1 | 3 | 1478 |