Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272036.1 | NP_001258965.1 | 5 | 1639 | ||
NM_014665.3 | NP_055480.1 | 4 | 1572 | |||
XM_005272358.4 | XP_005272415.1 | 4 | 1480 | |||
XM_005272359.4 | XP_005272416.1 | 5 | 1519 | |||
XM_005272360.4 | XP_005272417.1 | 4 | 1346 | |||
XM_017014005.1 | XP_016869494.1 | 5 | 1386 | |||
GenBank mRNA | AK289473.1 | - | 4 | 1540 | ||
BC011377.1 | - | 5 | 1599 | 3853673 | ||
D25216.1 | - | 4 | 1530 |