Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195555.1 | NP_001182484.1 | 6 | 842 | ||
NM_001195556.1 | NP_001182485.1 | 6 | 819 | |||
NM_014666.3 | NP_055481.1 | 6 | 842 | |||
XM_011534701.2 | XP_011533003.1 | 6 | 827 | |||
XM_017010087.1 | XP_016865576.1 | 6 | 827 | |||
XM_017010088.1 | XP_016865577.1 | 6 | 827 | |||
GenBank mRNA | AF434813.1 | - | 6 | 637 | ||
AK092765.1 | - | 6 | 789 | |||
AK300257.1 | - | 6 | 810 | |||
AL529948.3 | - | 6 | 829 | |||
BC004467.2 | - | 6 | 703 | 3831533 | ||
D79993.1 | - | 6 | 738 | |||
KU178486.1 | - | 6 | 637 | |||
KU178487.1 | - | 6 | 637 |