Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrb1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085403.1 | NP_001078872.1 | 3 | 248 | ||
XM_008763329.1 | XP_008761551.1 | 4 | 358 | |||
XM_008763330.1 | XP_008761552.1 | 4 | 631 | |||
XM_008763331.1 | XP_008761553.1 | 2 | 650 | |||
GenBank mRNA | EF100679.1 | - | 3 | 248 | ||
EF100685.1 | - | 3 | 248 | |||
X97477.1 | - | 3 | 248 |