Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST18 SIRNA Assays ›
Gene Symbol:
ST18
Gene Name:
ST18, C2H2C-type zinc finger
Gene Aliases:
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location:
Chr.8: 52110832 - 52409879 on Build GRCh38
UniGene ID:
Hs.655499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18701
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014682.2
NP_055497.1
16
2825
XM_006716487.1
XP_006716550.1
16
2592
XM_011517629.1
XP_011515931.1
17
2748
XM_011517631.1
XP_011515933.1
18
2789
XM_011517632.1
XP_011515934.1
16
2603
XM_011517633.1
XP_011515935.1
16
2596
XM_011517634.1
XP_011515936.1
16
2792
XM_011517635.1
XP_011515937.1
16
2796
XM_011517636.2
XP_011515938.1
15
2873
XM_011517637.1
XP_011515939.1
14
2330
XM_011517638.2
XP_011515940.1
13
2371
XM_011517641.1
XP_011515943.1
18
2836
XM_017014047.1
XP_016869536.1
16
2952
XM_017014048.1
XP_016869537.1
16
2956
XM_017014049.1
XP_016869538.1
17
3002
XM_017014050.1
XP_016869539.1
16
2789
XM_017014051.1
XP_016869540.1
15
2796
XM_017014052.1
XP_016869541.1
17
3006
XM_017014053.1
XP_016869542.1
17
2993
XM_017014054.1
XP_016869543.1
15
3042
XM_017014055.1
XP_016869544.1
15
2633
XM_017014056.1
XP_016869545.1
18
3043
XM_017014057.1
XP_016869546.1
16
2846
XM_017014058.1
XP_016869547.1
16
2607
XM_017014059.1
XP_016869548.1
16
2793
XM_017014060.1
XP_016869549.1
17
2830
XM_017014061.1
XP_016869550.1
15
2965
XM_017014062.1
XP_016869551.1
14
2717
XM_017014063.1
XP_016869552.1
15
3263
XM_017014064.1
XP_016869553.1
15
2877
XM_017014065.1
XP_016869554.1
17
2997
XM_017014066.1
XP_016869555.1
15
2636
XM_017014067.1
XP_016869556.1
15
3046
XM_017014068.1
XP_016869557.1
16
2837
XM_017014069.1
XP_016869558.1
13
2534
XM_017014070.1
XP_016869559.1
12
2455
XM_017014071.1
XP_016869560.1
13
2703
XM_017014072.1
XP_016869561.1
17
2867
XM_017014073.1
XP_016869562.1
17
2505
XM_017014074.1
XP_016869563.1
16
2788
XM_017014075.1
XP_016869564.1
17
3039
XM_017014076.1
XP_016869565.1
17
3043
XM_017014077.1
XP_016869566.1
17
2680
XM_017014078.1
XP_016869567.1
16
2960
XM_017014079.1
XP_016869568.1
16
3351
XM_017014080.1
XP_016869569.1
14
2592
GenBank mRNA
AB011107.1
-
16
2689
BC117147.1
-
11
2082
40125698
BC117149.1
-
11
2082
40125700
BC143872.1
-
11
2077
BC143873.1
-
12
2170
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0