Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST18 SIRNA Assays ›
Gene Symbol:
ST18
Gene Name:
ST18, C2H2C-type zinc finger
Gene Aliases:
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location:
Chr.8: 52110832 - 52409879 on Build GRCh38
UniGene ID:
Hs.655499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18703
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014682.2
NP_055497.1
25
3700
XM_006716487.1
XP_006716550.1
25
3467
XM_011517629.1
XP_011515931.1
26
3623
XM_011517631.1
XP_011515933.1
27
3664
XM_011517632.1
XP_011515934.1
25
3478
XM_011517633.1
XP_011515935.1
25
3471
XM_011517634.1
XP_011515936.1
25
3667
XM_011517635.1
XP_011515937.1
25
3671
XM_011517636.2
XP_011515938.1
24
3748
XM_011517637.1
XP_011515939.1
23
3205
XM_011517638.2
XP_011515940.1
22
3246
XM_011517641.1
XP_011515943.1
27
3711
XM_011517642.1
XP_011515944.1
25
3377
XM_017014047.1
XP_016869536.1
25
3827
XM_017014048.1
XP_016869537.1
25
3831
XM_017014049.1
XP_016869538.1
26
3877
XM_017014050.1
XP_016869539.1
25
3664
XM_017014051.1
XP_016869540.1
24
3671
XM_017014052.1
XP_016869541.1
26
3881
XM_017014053.1
XP_016869542.1
26
3868
XM_017014054.1
XP_016869543.1
24
3917
XM_017014055.1
XP_016869544.1
24
3508
XM_017014056.1
XP_016869545.1
27
3918
XM_017014057.1
XP_016869546.1
25
3721
XM_017014058.1
XP_016869547.1
25
3482
XM_017014059.1
XP_016869548.1
25
3668
XM_017014060.1
XP_016869549.1
26
3705
XM_017014061.1
XP_016869550.1
24
3840
XM_017014062.1
XP_016869551.1
23
3592
XM_017014063.1
XP_016869552.1
24
4138
XM_017014064.1
XP_016869553.1
24
3752
XM_017014065.1
XP_016869554.1
26
3872
XM_017014066.1
XP_016869555.1
24
3511
XM_017014067.1
XP_016869556.1
24
3921
XM_017014068.1
XP_016869557.1
25
3712
XM_017014069.1
XP_016869558.1
22
3409
XM_017014070.1
XP_016869559.1
21
3330
XM_017014071.1
XP_016869560.1
22
3578
XM_017014072.1
XP_016869561.1
26
3742
XM_017014073.1
XP_016869562.1
26
3380
XM_017014074.1
XP_016869563.1
25
3663
XM_017014075.1
XP_016869564.1
26
3914
XM_017014076.1
XP_016869565.1
26
3918
XM_017014077.1
XP_016869566.1
26
3555
XM_017014078.1
XP_016869567.1
25
3835
XM_017014079.1
XP_016869568.1
25
4226
XM_017014080.1
XP_016869569.1
23
3467
XM_017014081.1
XP_016869570.1
24
3588
XM_017014082.1
XP_016869571.1
24
3423
XM_017014083.1
XP_016869572.1
23
3263
XM_017014084.1
XP_016869573.1
21
3001
GenBank mRNA
AB011107.1
-
25
3564
BC117147.1
-
20
2957
40125698
BC117149.1
-
20
2957
40125700
BC143872.1
-
20
2952
BC143873.1
-
21
3045
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cs4xr:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0