Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001281.1 | NP_001001281.1 | 1 | 248 |
Gene Symbol: LOC684170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748937.2 | XP_003748985.1 | 1 | 248 |
Set Membership: |
![]() |