Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099513.1 | NP_001092983.1 | 2 | 376 |
Gene Symbol: LOC686837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769102.1 | XP_008767324.1 | 5 | 655 |
Set Membership: |
![]() |