Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012279.2 | NP_001012279.1 | 11 | 4770 | ||
GenBank mRNA | AK091416.1 | - | 8 | 2541 |
Gene Symbol: KIAA0408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014702.4 | NP_055517.3 | 5 | 1076 | ||
GenBank mRNA | AB007868.1 | - | 3 | 1469 | ||
AK091424.1 | - | 6 | 1432 | |||
AK125983.1 | - | 6 | 1585 | |||
BC018396.1 | - | 5 | 914 | 4184033 |