Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130508.1 | NP_001123980.1 | 2 | 994 | ||
GenBank mRNA | BC169036.1 | - | 1 | 1849 | 7105960 |