Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ddr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031764.3 | NP_113952.2 | 13 | 1975 | ||
XM_006250226.2 | XP_006250288.1 | 14 | 5272 | |||
XM_006250227.2 | XP_006250289.1 | 16 | 2607 | |||
XM_006250228.2 | XP_006250290.1 | 15 | 2388 | |||
XM_006250229.2 | XP_006250291.1 | 15 | 2336 | |||
XM_008769761.1 | XP_008767983.1 | 15 | 2350 | |||
XM_008769762.1 | XP_008767984.1 | 15 | 2282 | |||
XM_008769763.1 | XP_008767985.1 | 15 | 2588 | |||
GenBank mRNA | BC161961.1 | - | 13 | 1975 | 9027893 | |
EV765307.1 | - | 2 | 221 |