Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM94 SIRNA Assays ›
Gene Symbol:
TMEM94
Gene Name:
transmembrane protein 94
Gene Aliases:
KIAA0195
Chromosome Location:
Chr.17: 75441159 - 75500452 on Build GRCh38
UniGene ID:
Hs.514474
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18872
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM94
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321148.1
NP_001308077.1
10
1305
NM_001321149.1
NP_001308078.1
11
1477
NM_014738.5
NP_055553.3
11
1477
XM_005257864.2
XP_005257921.1
10
1300
XM_011525514.1
XP_011523816.1
11
1358
XM_011525515.1
XP_011523817.1
12
1484
XM_011525516.1
XP_011523818.1
11
1274
XM_011525517.1
XP_011523819.1
11
1253
XM_011525519.1
XP_011523821.1
5
414
XM_011525520.2
XP_011523822.1
8
914
XM_017025433.1
XP_016880922.1
10
1300
XM_017025434.1
XP_016880923.1
10
1300
XM_017025435.1
XP_016880924.1
10
1300
XM_017025436.1
XP_016880925.1
10
1300
XM_017025437.1
XP_016880926.1
10
1300
XM_017025438.1
XP_016880927.1
10
1300
XM_017025439.1
XP_016880928.1
10
1300
XM_017025440.1
XP_016880929.1
11
1358
XM_017025441.1
XP_016880930.1
10
1300
XM_017025442.1
XP_016880931.1
10
1300
XM_017025443.1
XP_016880932.1
10
1300
XM_017025444.1
XP_016880933.1
10
1300
XM_017025445.1
XP_016880934.1
10
1300
XM_017025446.1
XP_016880935.1
10
1300
XM_017025447.1
XP_016880936.1
10
1300
XM_017025448.1
XP_016880937.1
10
1300
XM_017025449.1
XP_016880938.1
11
1358
XM_017025450.1
XP_016880939.1
10
1300
GenBank mRNA
AK128508.1
-
3
152
AK294526.1
-
10
1268
AK304819.1
-
3
252
AK316060.1
-
10
1305
BC045540.1
-
11
1246
5274473
CR936809.1
-
9
1546
D83779.1
-
11
1286
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f2nf4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0