Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077440.1 | NP_001070908.1 | 9 | 2343 | ||
NM_001077441.1 | NP_001070909.1 | 9 | 1830 | |||
NM_001301038.1 | NP_001287967.1 | 9 | 2343 | |||
NM_014739.2 | NP_055554.1 | 9 | 2349 | |||
XM_005267237.3 | XP_005267294.1 | 9 | 2285 | |||
XM_017011529.1 | XP_016867018.1 | 9 | 1817 | |||
XM_017011530.1 | XP_016867019.1 | 9 | 2624 | |||
XM_017011531.1 | XP_016867020.1 | 9 | 1760 | |||
XM_017011532.1 | XP_016867021.1 | 10 | 2512 | |||
GenBank mRNA | AF249273.1 | - | 7 | 2090 | ||
AK308870.1 | - | 1 | 177 | |||
AK309259.1 | - | 1 | 400 | |||
BC063846.1 | - | 9 | 2256 | |||
BC132780.1 | - | 9 | 2293 | 40146802 | ||
BC144281.1 | - | 9 | 2287 | 9052801 | ||
BC144299.1 | - | 9 | 2293 | |||
BX648878.1 | - | 5 | 1090 | |||
BX649160.1 | - | 9 | 1761 | |||
D79986.1 | - | 9 | 2349 |