Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATG13 SIRNA Assays ›
Gene Symbol:
ATG13
Gene Name:
autophagy related 13
Gene Aliases:
KIAA0652, PARATARG8
Chromosome Location:
Chr.11: 46617276 - 46676019 on Build GRCh38
UniGene ID:
Hs.127403
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18880
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142673.2
NP_001136145.1
13, 14
1724
NM_001205119.1
NP_001192048.1
13, 14
1767
NM_001205120.1
NP_001192049.1
12, 13
1668
NM_001205121.1
NP_001192050.1
12, 13
1608
NM_001205122.1
NP_001192051.1
10, 11
1475
NM_014741.4
NP_055556.2
12, 13
1613
XM_005253262.2
XP_005253319.1
14, 15
1563
XM_005253263.2
XP_005253320.1
14, 15
1854
XM_005253265.2
XP_005253322.1
15, 16
2060
XM_005253266.2
XP_005253323.1
14, 15
1789
XM_005253268.2
XP_005253325.1
12, 13
1166
XM_006718394.2
XP_006718457.1
15, 16
1910
XM_006718395.2
XP_006718458.1
15, 16
1845
XM_006718396.2
XP_006718459.1
14, 15
1944
XM_011520493.1
XP_011518795.1
16, 17
2047
XM_011520496.2
XP_011518798.1
13, 14
1822
XM_011520499.2
XP_011518801.1
13, 14
1436
XM_011520500.1
XP_011518802.1
15, 16
1883
XM_011520501.1
XP_011518803.1
14, 15
1811
XM_011520502.1
XP_011518804.1
14, 15
1784
XM_011520503.1
XP_011518805.1
13, 14
1700
XM_017018599.1
XP_016874088.1
15, 16
1995
XM_017018600.1
XP_016874089.1
14, 15
1536
XM_017018601.1
XP_016874090.1
13, 14
1711
XM_017018602.1
XP_016874091.1
14, 15
1961
XM_017018603.1
XP_016874092.1
15, 16
2078
XM_017018604.1
XP_016874093.1
13, 14
1755
XM_017018605.1
XP_016874094.1
14, 15
1746
XM_017018606.1
XP_016874095.1
13, 14
1690
XM_017018607.1
XP_016874096.1
11, 12
1067
XM_017018608.1
XP_016874097.1
13, 14
1437
XM_017018609.1
XP_016874098.1
13, 14
1728
XM_017018610.1
XP_016874099.1
12, 13
1612
XM_017018611.1
XP_016874100.1
13, 14
1663
XM_017018612.1
XP_016874101.1
13, 14
1850
XM_017018613.1
XP_016874102.1
12, 13
1644
XM_017018614.1
XP_016874103.1
13, 14
1635
XM_017018615.1
XP_016874104.1
12, 13
1734
XM_017018616.1
XP_016874105.1
11, 12
1528
XM_017018617.1
XP_016874106.1
13, 14
1733
XM_017018618.1
XP_016874107.1
15, 16
2049
XM_017018619.1
XP_016874108.1
13, 14
1834
XM_017018620.1
XP_016874109.1
13, 14
1894
XM_017018621.1
XP_016874110.1
12, 13
1778
XM_017018622.1
XP_016874111.1
12, 13
1735
GenBank mRNA
AB014552.1
-
12, 13
1348
AK023867.1
-
12, 13
1346
AK223603.1
-
13, 14
1443
AK289641.1
-
13, 14
1271
AK293109.1
-
12, 13
1312
AK294110.1
-
10, 11
1188
AK296526.1
-
5, 6
555
AK315925.1
-
5, 6
482
BC001331.1
-
12, 13
1042
3458359
BC002378.2
-
12, 13
1312
2961127
BC006191.2
-
12, 13
1354
3936851
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
macroautophagy
mitophagy in response to mitochondrial depolarization
positive regulation of protein targeting to mitochondrion
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline