Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134702.1 | NP_001128174.1 | 3 | 407 | ||
XM_006251529.2 | XP_006251591.1 | 3 | 1655 | |||
GenBank mRNA | BC161984.1 | - | 3 | 407 | 9030600 |