Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Naa16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751517.3 | XP_003751565.1 | 3 | 351 | ||
XM_008770925.1 | XP_008769147.1 | 3 | 351 | |||
XM_008770926.1 | XP_008769148.1 | 3 | 351 |