Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARMCX2 SIRNA Assays ›
Gene Symbol:
ARMCX2
Gene Name:
armadillo repeat containing, X-linked 2
Gene Aliases:
ALEX2, GASP9
Chromosome Location:
Chr.X: 101655281 - 101659891 on Build GRCh38
UniGene ID:
Hs.48924
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18996
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARMCX2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282231.1
NP_001269160.1
6
2219
NM_014782.6
NP_055597.1
5
2180
NM_177949.3
NP_808818.1
6
2231
XM_005278109.1
XP_005278166.1
6
2229
XM_005278110.1
XP_005278167.1
6
2262
XM_005278111.1
XP_005278168.1
6
2231
XM_005278113.1
XP_005278170.1
6
2206
XM_005278114.1
XP_005278171.1
6
2250
XM_005278115.1
XP_005278172.1
6
2219
XM_005278116.1
XP_005278173.1
6
2217
XM_005278117.1
XP_005278174.1
6
2187
XM_011531071.1
XP_011529373.1
6
2306
XM_011531072.1
XP_011529374.1
6
2273
XM_017029987.1
XP_016885476.1
3
1976
XM_017029988.1
XP_016885477.1
3
1973
XM_017029989.1
XP_016885478.1
6
2223
XM_017029990.1
XP_016885479.1
6
2261
XM_017029991.1
XP_016885480.1
5
2153
XM_017029992.1
XP_016885481.1
6
2242
XM_017029993.1
XP_016885482.1
5
2120
XM_017029994.1
XP_016885483.1
5
2108
XM_017029995.1
XP_016885484.1
4
2041
XM_017029996.1
XP_016885485.1
3
1951
XM_017029997.1
XP_016885486.1
2
1888
GenBank mRNA
AB011084.1
-
6
2177
AK291342.1
-
6
2190
BC012541.1
-
5
2136
4043663
BC015926.1
-
3
2034
3922790
BX648494.1
-
6
2178
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0