Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272011.1 | NP_001258940.1 | 3 | 386 | ||
NM_014226.2 | NP_055041.1 | 4 | 476 | |||
XM_006720225.2 | XP_006720288.1 | 4 | 476 | |||
XM_006720226.2 | XP_006720289.1 | 4 | 476 | |||
XM_011537052.1 | XP_011535354.1 | 4 | 476 | |||
XM_017021549.1 | XP_016877038.1 | 4 | 476 | |||
XM_017021550.1 | XP_016877039.1 | 3 | 375 | |||
XM_017021551.1 | XP_016877040.1 | 5 | 416 | |||
XM_017021552.1 | XP_016877041.1 | 4 | 517 | |||
XM_017021555.1 | XP_016877044.1 | 4 | 476 | |||
XM_017021556.1 | XP_016877045.1 | 4 | 476 | |||
XM_017021557.1 | XP_016877046.1 | 3 | 376 | |||
XM_017021558.1 | XP_016877047.1 | 3 | 342 | |||
GenBank mRNA | AB022694.1 | - | 4 | 470 | ||
AK302349.1 | - | 3 | 371 | |||
AK312778.1 | - | 4 | 467 | |||
BC026069.1 | - | 4 | 466 | 4838296 | ||
BC043179.1 | - | 1 | 1068 | 5287879 | ||
BC053536.1 | - | 4 | 452 | 5175851 | ||
DB037795.1 | - | 4 | 476 |