Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109606.1 | NP_001103076.1 | 8, 9 | 962 | ||
XM_006234656.2 | XP_006234718.1 | 8, 9 | 987 | |||
XM_006234657.2 | XP_006234719.1 | 8, 9 | 987 | |||
GenBank mRNA | FQ210238.1 | - | 1, 2 | 105 |