Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atp5j2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271117.1 | NP_001258046.1 | 3 | 214 | ||
GenBank mRNA | FQ210117.1 | - | 3 | 201 | ||
FQ211282.1 | - | 3 | 201 | |||
FQ216873.1 | - | 3 | 189 | |||
FQ217676.1 | - | 3 | 189 | |||
FQ218321.1 | - | 3 | 203 | |||
FQ221351.1 | - | 3 | 193 | |||
FQ221691.1 | - | 3 | 201 | |||
FQ221732.1 | - | 3 | 183 | |||
FQ222492.1 | - | 3 | 188 | |||
FQ223220.1 | - | 3 | 188 | |||
FQ223295.1 | - | 3 | 191 | |||
FQ223575.1 | - | 3 | 186 | |||
FQ223738.1 | - | 3 | 189 | |||
FQ223886.1 | - | 2 | 167 | |||
FQ223939.1 | - | 3 | 189 | |||
FQ224192.1 | - | 3 | 189 | |||
FQ224234.1 | - | 3 | 203 | |||
FQ224423.1 | - | 3 | 201 | |||
FQ224479.1 | - | 3 | 201 | |||
FQ224602.1 | - | 3 | 202 | |||
FQ224807.1 | - | 3 | 201 | |||
FQ228438.1 | - | 3 | 189 |
Gene Symbol: LOC689271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001070212.4 | XP_001070212.1 | 1 | 181 |