Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam101a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109547.1 | NP_001103017.1 | 3 | 431 | ||
XM_008769261.1 | XP_008767483.1 | 3 | 181 | |||
XM_008769262.1 | XP_008767484.1 | 3 | 767 | |||
GenBank mRNA | FQ235329.1 | - | 3 | 220 |