Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039459.2 | NP_001034548.1 | 6 | 697 | ||
NM_001206480.2 | NP_001193409.1 | 21 | 2476 | |||
NM_001206482.1 | NP_001193411.1 | 21 | 2646 | |||
NM_014800.10 | NP_055615.8 | 21 | 2287 | |||
NM_130442.3 | NP_569709.1 | 6 | 843 | |||
NR_038120.1 | 5 | 730 | ||||
XM_005249919.2 | XP_005249976.1 | 22 | 2840 | |||
XM_006715805.1 | XP_006715868.1 | 21 | 2132 | |||
XM_011515654.1 | XP_011513956.1 | 22 | 3525 | |||
XM_017012838.1 | XP_016868327.1 | 22 | 3710 | |||
XM_017012839.1 | XP_016868328.1 | 21 | 2074 | |||
GenBank mRNA | AF398885.1 | - | 20 | 1938 | ||
AK126565.1 | - | 9 | 1327 | |||
AK291784.1 | - | 21 | 2646 | |||
AK296253.1 | - | 18 | 1807 | |||
AK301151.1 | - | 5 | 605 | |||
AK314349.1 | - | 21 | 2076 | |||
AL136787.1 | - | 21 | 2188 | |||
BC003051.1 | - | 6 | 693 | |||
BC064635.1 | - | 6 | 645 | |||
BC077074.1 | - | 6 | 694 | |||
BC114341.2 | - | 21 | 2025 | 40035461 | ||
D87457.1 | - | 6 | 669 |