Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039459.2 | NP_001034548.1 | 2, 3 | 360 | ||
NM_001206480.2 | NP_001193409.1 | 17, 18 | 2139 | |||
NM_001206482.1 | NP_001193411.1 | 17, 18 | 2309 | |||
NM_014800.10 | NP_055615.8 | 17, 18 | 1950 | |||
NM_130442.3 | NP_569709.1 | 2, 3 | 506 | |||
XM_005249919.2 | XP_005249976.1 | 18, 19 | 2503 | |||
XM_006715805.1 | XP_006715868.1 | 17, 18 | 1795 | |||
XM_011515654.1 | XP_011513956.1 | 18, 19 | 3188 | |||
XM_017012838.1 | XP_016868327.1 | 18, 19 | 3373 | |||
XM_017012839.1 | XP_016868328.1 | 17, 18 | 1737 | |||
GenBank mRNA | AF398885.1 | - | 16, 17 | 1601 | ||
AK126565.1 | - | 5, 6 | 990 | |||
AK291784.1 | - | 17, 18 | 2309 | |||
AK296253.1 | - | 14, 15 | 1470 | |||
AK314349.1 | - | 17, 18 | 1739 | |||
AL136787.1 | - | 17, 18 | 1851 | |||
BC003051.1 | - | 2, 3 | 356 | |||
BC064635.1 | - | 2, 3 | 308 | |||
BC077074.1 | - | 2, 3 | 357 | |||
BC114341.2 | - | 17, 18 | 1688 | 40035461 | ||
BI600726.1 | - | 2, 3 | 368 | |||
D87457.1 | - | 2, 3 | 332 |