Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134799.1 | NP_001128271.1 | 2 | 180 | ||
XM_006246813.2 | XP_006246875.1 | 1, 2 | 336 | |||
XM_008767876.1 | XP_008766098.1 | 1, 2 | 336 | |||
XM_008767877.1 | XP_008766099.1 | 1, 2 | 336 | |||
XM_008767878.1 | XP_008766100.1 | 1, 2 | 336 |
Gene Symbol: Kctd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC168958.1 | - | 642 |