Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm20l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001072851.4 | XP_001072851.3 | 4 | 823 | ||
XM_008764723.1 | XP_008762945.1 | 5 | 952 |