Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP170 SIRNA Assays ›
Gene Symbol:
CEP170
CEP170P1
Gene Name:
centrosomal protein 170
centrosomal protein 170 pseudogene 1
Gene Aliases:
FAM68A, KAB, KIAA0470
CEP170L, FAM68B, KIAA0470L
Chromosome Location:
Chr.1: 243124428 - 243255936 on Build GRCh38
Chromosome Location:
Chr.4: 118516340 - 118554204 on Build GRCh38
UniGene ID:
Hs.533635
Hs.408293
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s19088
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP170
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042404.1
NP_001035863.1
15
4243
NM_001042405.1
NP_001035864.1
15
4165
NM_014812.2
NP_055627.2
16
4537
XM_006711843.3
XP_006711906.1
15
5276
XM_011544334.2
XP_011542636.1
15
5283
XM_011544335.2
XP_011542637.1
15
4696
XM_011544336.2
XP_011542638.1
15
5602
XM_011544337.2
XP_011542639.1
15
5253
XM_011544338.2
XP_011542640.1
15
5202
XM_011544339.2
XP_011542641.1
15
5175
XM_011544340.2
XP_011542642.1
15
5095
XM_011544341.2
XP_011542643.1
16
5047
XM_011544342.2
XP_011542644.1
15
5044
XM_011544343.2
XP_011542645.1
14
4756
XM_011544344.2
XP_011542646.1
15
4803
XM_017002932.1
XP_016858421.1
15
5196
XM_017002933.1
XP_016858422.1
16
4867
XM_017002934.1
XP_016858423.1
16
4510
XM_017002935.1
XP_016858424.1
16
4966
XM_017002936.1
XP_016858425.1
16
4480
XM_017002937.1
XP_016858426.1
16
4939
XM_017002938.1
XP_016858427.1
16
4909
XM_017002939.1
XP_016858428.1
16
4786
XM_017002940.1
XP_016858429.1
16
4429
XM_017002941.1
XP_016858430.1
16
4759
XM_017002942.1
XP_016858431.1
16
4858
XM_017002943.1
XP_016858432.1
16
4678
XM_017002944.1
XP_016858433.1
15
4796
XM_017002945.1
XP_016858434.1
15
4722
XM_017002946.1
XP_016858435.1
15
4570
XM_017002947.1
XP_016858436.1
15
4665
XM_017002948.1
XP_016858437.1
15
4658
XM_017002949.1
XP_016858438.1
15
4614
XM_017002950.1
XP_016858439.1
15
4489
XM_017002951.1
XP_016858440.1
14
4276
GenBank mRNA
AB007939.1
-
14
3810
AB022657.1
-
15
4182
AB022658.1
-
14
3888
AB022659.1
-
14
3810
AK001664.1
-
4
1825
AK313722.1
-
3
327
AL161964.1
-
1
326
BC078155.1
-
4
782
BC140794.1
-
14
3879
9021662
BC146790.1
-
14
3810
9007254
Gene Symbol: CEP170P1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_003135.2
4
340
GenBank mRNA
BC014590.2
-
4
340
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Process(es)
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0