Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PJA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014819.4 | NP_055634.3 | 4 | 808 | ||
XM_017010099.1 | XP_016865588.1 | 5 | 884 | |||
GenBank mRNA | AB007898.1 | - | 4 | 705 | ||
AK291759.1 | - | 4 | 742 | |||
BC030826.1 | - | 4 | 704 | 4811738 | ||
BQ228553.1 | - | 4 | 715 |