Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166577.1 | NP_001160049.1 | 2 | 238 | ||
XM_008768394.1 | XP_008766616.1 | 3 | 564 | |||
XM_008768395.1 | XP_008766617.1 | 3 | 568 | |||
GenBank mRNA | GU057983.1 | - | 2 | 238 |