Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam109b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130511.1 | NP_001123983.1 | 2 | 1041 | ||
XM_006242124.2 | XP_006242186.1 | 2 | 1627 | |||
XM_006242125.2 | XP_006242187.1 | 2 | 1608 | |||
XM_006242126.2 | XP_006242188.1 | 2 | 1395 |