Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNAP91 SIRNA Assays ›
Gene Symbol:
SNAP91
Gene Name:
synaptosome associated protein 91
Gene Aliases:
AP180, CALM
Chromosome Location:
Chr.6: 83552885 - 83709408 on Build GRCh38
UniGene ID:
Hs.368046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19162
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNAP91
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242792.1
NP_001229721.1
8
1033
NM_001242793.1
NP_001229722.1
8
832
NM_001242794.1
NP_001229723.1
8
832
NM_001256717.1
NP_001243646.1
11
1268
NM_001256718.1
NP_001243647.1
7
743
NM_014841.2
NP_055656.1
8
1036
XM_005248770.4
XP_005248827.1
8
875
XM_006715615.1
XP_006715678.1
8
827
XM_011536265.1
XP_011534567.1
8
834
XM_011536266.1
XP_011534568.1
8
946
XM_011536269.1
XP_011534571.1
8
827
XM_011536271.1
XP_011534573.1
8
827
XM_011536273.2
XP_011534575.1
8
827
XM_011536275.2
XP_011534577.1
8
827
XM_011536276.2
XP_011534578.1
8
827
XM_011536277.1
XP_011534579.1
10
1115
XM_017011554.1
XP_016867043.1
8
991
XM_017011555.1
XP_016867044.1
8
875
XM_017011556.1
XP_016867045.1
8
807
XM_017011557.1
XP_016867046.1
8
1004
XM_017011558.1
XP_016867047.1
8
834
XM_017011559.1
XP_016867048.1
8
933
XM_017011560.1
XP_016867049.1
8
875
XM_017011561.1
XP_016867050.1
8
946
XM_017011562.1
XP_016867051.1
8
1004
XM_017011563.1
XP_016867052.1
8
1004
XM_017011564.1
XP_016867053.1
8
875
XM_017011565.1
XP_016867054.1
8
933
XM_017011566.1
XP_016867055.1
8
1004
XM_017011567.1
XP_016867056.1
8
807
XM_017011568.1
XP_016867057.1
8
1004
XM_017011569.1
XP_016867058.1
8
1004
XM_017011570.1
XP_016867059.1
8
1004
XM_017011571.1
XP_016867060.1
8
875
XM_017011572.1
XP_016867061.1
8
834
XM_017011573.1
XP_016867062.1
8
933
XM_017011574.1
XP_016867063.1
8
1004
XM_017011575.1
XP_016867064.1
8
1004
XM_017011576.1
XP_016867065.1
8
807
XM_017011577.1
XP_016867066.1
8
933
XM_017011578.1
XP_016867067.1
8
946
XM_017011579.1
XP_016867068.1
8
1004
XM_017011580.1
XP_016867069.1
8
1004
XM_017011581.1
XP_016867070.1
8
834
XM_017011582.1
XP_016867071.1
8
863
XM_017011583.1
XP_016867072.1
8
991
XM_017011584.1
XP_016867073.1
8
834
XM_017011585.1
XP_016867074.1
8
875
XM_017011586.1
XP_016867075.1
8
827
XM_017011587.1
XP_016867076.1
8
933
XM_017011588.1
XP_016867077.1
8
946
XM_017011589.1
XP_016867078.1
8
1004
XM_017011590.1
XP_016867079.1
8
875
GenBank mRNA
AB014556.1
-
8
962
AK289582.1
-
8
1004
AK294898.1
-
7
727
BC060818.1
-
8
816
4827211
CR749348.1
-
11
1235
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031728, XM_008766455, XM_006243494, XM_008766456, XM_006243497, and others.
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
protein transport
clathrin coat assembly
regulation of clathrin-mediated endocytosis
protein binding
1-phosphatidylinositol binding
protein kinase binding
clathrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0